missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethylthio)benzoic acid, 97%

Product Code. 11418857
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418857 5 g 5g
11408857 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418857 Supplier Thermo Scientific Alfa Aesar Supplier No. A19975.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 946-65-6 | C8H4F3O2S | 221.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 946-65-6
Molecular Formula C8H4F3O2S
Molecular Weight (g/mol) 221.17
MDL Number MFCD00236337
InChI Key IVGAPIVNQABETQ-UHFFFAOYSA-M
Synonym 3-trifluoromethylthio benzoic acid, 3-trifluoromethyl thio benzoic acid, 3-trifluoromethyl sulfanyl benzoic acid, 3-trifluoromethylsulfanyl benzoic acid, 3-trifluoromethylsulfanyl-benzoic acid, benzoic acid, 3-trifluoromethyl thio, 3-trifluoromethyl sulphanyl benzoic acid, 3-trifluoromethylthio benzoicacid, 3-carboxyphenyl trifluoromethyl sulphide
PubChem CID 688330
IUPAC Name 3-(trifluoromethylsulfanyl)benzoic acid
SMILES [O-]C(=O)C1=CC=CC(SC(F)(F)F)=C1

Specifications

Melting Point 71°C to 74°C
Quantity 5 g
Formula Weight 222.18
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethylthio)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.