missing translation for 'onlineSavingsMsg'
Learn More

3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one, 97%, Thermo Scientific™

Product Code. 11976021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11976021 1 g 1g
11986021 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976021 Supplier Thermo Scientific Acros Supplier No. 440990010
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket
This item is not returnable. View return policy
for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34932-07-5
Molecular Formula C22H19O2P
Molecular Weight (g/mol) 346.37
MDL Number MFCD00065318
InChI Key JUKRJGQYUNADDD-UHFFFAOYSA-N
Synonym 2 3h-furanone, 3-triphenylphosphoranylidene, gamma-butyrolactonylidene triphenyl phosphorane, 2-triphenylphosphoranylidene-gamma-butyrolactone, alpha-triphenylphosphoranylidene-gamma-butyrolactone, 2 3h-furanone, dihydro-3-triphenylphosphoranylidene, 3-triphenylphosphoranylidene dihydro-2 3h-furanone, 3-triphenyl-??-phosphanylidene oxolan-2-one, 3-triphenyl-, e5-phosphanylidene dihydrofuran-2 3h-one, .gamma.-butyrolactonylidene triphenyl phosphorane, 2-oxotetrahydrofuran-3-ylidenetriphenylphosphorane
PubChem CID 97111
IUPAC Name 3-(triphenyl-$l^{5}-phosphanylidene)oxolan-2-one
SMILES O=C1OCCC1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 243.0°C to 246.0°C
Color White
Elemental Analysis 73.24 to 79.34% (C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 346.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.