missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyl(methyl)amino)-N-methoxy-N-methylbenzamide, TRC

Product Code. 30335113
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335113 50 mg 50mg
30335689 10 mg 10mg
30346696 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30335113 Supplier Toronto Research Chemicals Supplier No. TRCB28785550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Benzyl(methyl)amino)-N-methoxy-N-methylbenzamide
Analyte or Component Names 4-(Benzyl(methyl)amino)-N-methoxy-N-methylbenzamide
Shipping Condition Room Temperature
Molecular Formula C17 H20 N2 O2
InChI Formula InChI=1S/C17H20N2O2/c1-18(13-14-7-5-4-6-8-14)16-11-9-15(10-12-16)17(20)19(2)21-3/h4-12H,13H2,1-3H3
Solution Type Neat
SMILES CON(C)C(=O)c1ccc(cc1)N(C)Cc2ccccc2
IUPAC Name 4-[benzyl(methyl)amino]-N-methoxy-N-methylbenzamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 284.353
Quantity 50 mg
Formula Weight 284.152
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.