missing translation for 'onlineSavingsMsg'
Learn More

2-(((2-Amino-5-nitrophenyl)(2-chlorophenyl)methylene)amino)acetic Acid, TRC

Product Code. 30338906
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30338906 100 mg
30335459 10 mg
30338367 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30338906 Supplier Toronto Research Chemicals Supplier No. TRCA997140100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(((2-Amino-5-nitrophenyl)(2-chlorophenyl)methylene)amino)acetic Acid
Analyte or Component Names 2-(((2-Amino-5-nitrophenyl)(2-chlorophenyl)methylene)amino)acetic Acid
Shipping Condition Room Temperature
Molecular Formula C15H12ClN3O4
InChI Formula InChI=1S/C15H12ClN3O4/c16-12-4-2-1-3-10(12)15(18-8-14(20)21)11-7-9(19(22)23)5-6-13(11)17/h1-7H,8,17H2,(H,20,21)
Solution Type Neat
SMILES ClC1=CC=CC=C1/C(C2=C(N)C=CC([N+]([O-])=O)=C2)=N\CC(O)=O
IUPAC Name 2-[[(2-amino-5-nitrophenyl)-(2-chlorophenyl)methylidene]amino]acetic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 333.73
Quantity 100 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.