missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-chloropentanamide, TRC

Product Code. 30339893
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30339893 25 mg 25mg
30331280 50 mg 50mg
30342387 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30339893 Supplier Toronto Research Chemicals Supplier No. TRCB23363525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyl-4-chloropentanamide
Analyte or Component Names N-Benzyl-4-chloropentanamide
Shipping Condition Room Temperature
Molecular Formula C3 H8 Br O3 P
InChI Formula InChI=1S/C3H8BrO3P/c4-2-1-3-8(5,6)7/h1-3H2,(H2,5,6,7)
Solution Type Neat
SMILES OP(=O)(O)CCCBr
IUPAC Name 3-bromopropylphosphonic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 202.972
Quantity 25 mg
Formula Weight 201.939
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.