missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzo[d][1,3]dioxol-5-yl)-N-benzyl-N,2-dimethylpropan-1-amine, TRC

Product Code. 30343622
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343622 250 mg 250mg
30330431 500 mg 500mg
30346194 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30343622 Supplier Toronto Research Chemicals Supplier No. TRCB857565250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Benzo[d][1,3]dioxol-5-yl)-N-benzyl-N,2-dimethylpropan-1-amine
Analyte or Component Names 3-(Benzo[d][1,3]dioxol-5-yl)-N-benzyl-N,2-dimethylpropan-1-amine
Shipping Condition Room Temperature
Molecular Formula C19 H23 N O2
InChI Formula InChI=1S/C19H23NO2/c1-15(12-20(2)13-16-6-4-3-5-7-16)10-17-8-9-18-19(11-17)22-14-21-18/h3-9,11,15H,10,12-14H2,1-2H3
Solution Type Neat
SMILES CC(CN(C)Cc1ccccc1)Cc2ccc3OCOc3c2
IUPAC Name 3-(1,3-benzodioxol-5-yl)-N-benzyl-N,2-dimethylpropan-1-amine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 297.391
Quantity 250 mg
Formula Weight 297.173
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.