missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(3-(Benzyloxy)-2,2-dimethylpropoxy)-6-oxohexanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 6-(3-(Benzyloxy)-2,2-dimethylpropoxy)-6-oxohexanoic Acid |
| Analyte or Component Names | 6-(3-(Benzyloxy)-2,2-dimethylpropoxy)-6-oxohexanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H26 O5 |
| InChI Formula | InChI=1S/C18H26O5/c1-18(2,13-22-12-15-8-4-3-5-9-15)14-23-17(21)11-7-6-10-16(19)20/h3-5,8-9H,6-7,10-14H2,1-2H3,(H,19,20) |
| Solution Type | Neat |
| SMILES | CC(C)(COCc1ccccc1)COC(=O)CCCCC(=O)O |
| IUPAC Name | 6-(2,2-dimethyl-3-phenylmethoxypropoxy)-6-oxohexanoic acid |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 322.396 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?