missing translation for 'onlineSavingsMsg'
Learn More

CDDO Imidazolide, TRC

Product Code. 30349419
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349419 10 mg -
30368198 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30349419 Supplier Toronto Research Chemicals Supplier No. TRCC22809010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443104-02-7
Molecular Formula C34H43N3O3
Molecular Weight (g/mol) 541.72
Synonym 1-(2-Cyano-3,12,28-trioxooleana-1,9(11)-dien-28-yl)-1H-imidazole, 2-Cyano-3,12-dioxooleana-1,9(11)-dien-28-oic acid imidazolide, CDDO-Im, RTA 403, TP 235
IUPAC Name (4aR,6aR,6aS,6bR,8aS,12aS,14bS)-8a-(imidazole-1-carbonyl)-4,4,6a,6b,11,11,14b-heptamethyl-3,13-dioxo-4a,5,6,6a,7,8,9,10,12,12a-decahydropicene-2-carbonitrile
SMILES CC1(C)CC[C@@]2(CC[C@]3(C)[C@@H]([C@@H]2C1)C(=O)C=C4[C@@]5(C)C=C(C#N)C(=O)C(C)(C)[C@@H]5CC[C@]43C)C(=O)N6C=CN=C6

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C34H43N3O3/c1-29(2)10-12-34(28(40)37-15-14-36-20-37)13-11-33(7)26(22(34)18-29)23(38)16-25-31(5)17-21(19-35)27(39)30(3,4)24(31)8-9-32(25,33)6/h14-17,20,22,24,26H,8-13,18H2,1-7H3/t22-,24-,26-,31-,32+,33+,34-/m0/s1
Chemical Name or Material CDDO Imidazolide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.