missing translation for 'onlineSavingsMsg'
Learn More

Chlorthalidone Dimer Sodium Salt, TRC

Product Code. 30350870
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350870 1 mg 1mg
30355620 5 mg 5mg
30356798 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350870 Supplier Toronto Research Chemicals Supplier No. TRCC4375101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorthalidone Dimer Sodium Salt
Analyte or Component Names Chlorthalidone Dimer Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C28H19Cl2N3O8S2.x(Na)
Synonym 2-Chloro-N-(2-chloro-5-(1-hydroxy-3-oxoisoindolin-1-yl)phenylsulfonyl)-5-(1-hydroxy-3-oxoisoindolin-1-yl)benzenesulfonamide Sodium Salt
InChI Formula InChI=1S/C28H19Cl2N3O8S2/c29-21-11-9-15(27(36)19-7-3-1-5-17(19)25(34)31-27)13-23(21)42(38,39)33-43(40,41)24-14-16(10-12-22(24)30)28(37)20-8-4-2-6-18(20)26(35)32-28/h1-14,33,36-37H,(H,31,34)(H,32,35)
Solution Type Neat
SMILES ClC1=C(S(NS(C2=C(Cl)C=CC(C3(O)C4=CC=CC=C4C(O)=N3)=C2)(=O)=O)(=O)=O)C=C(C5(O)C6=CC=CC=C6C(O)=N5)C=C1
IUPAC Name 2-chloro-N-[2-chloro-5-(1-hydroxy-3-oxo-2H-isoindol-1-yl)phenyl]sulfonyl-5-(1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 660.5
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.