missing translation for 'onlineSavingsMsg'
Learn More

Dehydrocorydaline nitrate, TRC

Product Code. 30351775
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351775 2.5 mg 2.5mg
30363593 10 mg 10mg
30351513 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351775 Supplier Toronto Research Chemicals Supplier No. TRCD2301232.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13005-09-9
Molecular Formula C22H24N2O7
Molecular Weight (g/mol) 428.4
Synonym 6H-Dibenzo[a,g]quinolizinium, 5-hydro-2,3,9,10-tetramethoxy-13-methyl-, nitrate (1:1) (ACI), Dibenzo[a,g]quinolizinium, 5,6-dihydro-2,3,9,10-tetramethoxy-13-methyl-, nitrate (9CI), Dehydrocorydaline nitrate, 5-Hydro-2,3,9,10-tetramethoxy-13-methyl-6H-dibenzo[a,g]quinolizinium nitrate
SMILES COc1cc2CC[n+]3cc4c(OC)c(OC)ccc4c(C)c3c2cc1OC.[O-][N+](=O)[O-]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C22H24NO4.NO3/c1-13-15-6-7-18(24-2)22(27-5)17(15)12-23-9-8-14-10-19(25-3)20(26-4)11-16(14)21(13)23;2-1(3)4/h6-7,10-12H,8-9H2,1-5H3;/q+1;-1
Formula Weight 428.1584
Chemical Name or Material Dehydrocorydaline nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.