missing translation for 'onlineSavingsMsg'
Learn More

9,13b-Dehydro Epinastine, TRC

Product Code. 30353393
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353393 25 mg 25mg
30361152 10 mg 10mg
30368725 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353393 Supplier Toronto Research Chemicals Supplier No. TRCD22960025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 706753-75-5
Molecular Formula C16 H13 N3
Molecular Weight (g/mol) 247.29
Synonym 9H-Dibenz[c,f]imidazo[1,5-a]azepin-3-amine, 9,13b-Dehydro Epinastine, 9,13b-Dehydroepinastine
SMILES Nc1ncc2c3ccccc3Cc4ccccc4n12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H13N3/c17-16-18-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)19(15)16/h1-8,10H,9H2,(H2,17,18)
Formula Weight 247.1109
Chemical Name or Material 9,13b-Dehydro Epinastine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.