missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol Caprylate, TRC

Product Code. 30354207
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354207 5 g 5g
30357664 25 g 25g
30357718 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354207 Supplier Toronto Research Chemicals Supplier No. TRCC2927505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1182-42-9
Molecular Formula C35 H60 O2
Molecular Weight (g/mol) 512.85
Synonym (3β)-Cholest-5-en-3-ol Octanoate, Cholesterol Octanoate, 3β-(Octanoyloxy)cholest-5-ene, 5-Cholesten-3β-ol Caprylate, Caprylic Acid Cholesterol Ester, Cholest-5-en-3β-ol caprylate, Cholesteryl Caprylate, Cholesteryl Octanoate
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] octanoate
SMILES CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C35H60O2/c1-7-8-9-10-11-15-33(36)37-28-20-22-34(5)27(24-28)16-17-29-31-19-18-30(26(4)14-12-13-25(2)3)35(31,6)23-21-32(29)34/h16,25-26,28-32H,7-15,17-24H2,1-6H3/t26-,28+,29+,30-,31+,32+,34+,35-/m1/s1
Formula Weight 512.4593
Chemical Name or Material Cholesteryl Caprylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.