missing translation for 'onlineSavingsMsg'
Learn More

Dibenzylfluorescein, TRC

Product Code. 30354695
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354695 10 mg 10mg
30362460 5 mg 5mg
30368430 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354695 Supplier Toronto Research Chemicals Supplier No. TRCD41740510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97744-44-0
Molecular Formula C34 H24 O5
Molecular Weight (g/mol) 512.551
Synonym 2-[3-Oxo-6-(phenylmethoxy)-3H-xanthen-9-yl]benzoic Acid Phenylmethyl Ester, NSC 645658
IUPAC Name 6,7-dibenzyl-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5c(Cc6ccccc6)c(Cc7ccccc7)ccc45)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C34H24O5/c35-24-12-15-27-30(19-24)38-31-20-25(36)13-16-28(31)34(27)29-14-11-23(17-21-7-3-1-4-8-21)26(32(29)33(37)39-34)18-22-9-5-2-6-10-22/h1-16,19-20,35-36H,17-18H2
Formula Weight 512.162
Chemical Name or Material Dibenzylfluorescein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.