missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Desmethyl Benoxaprofen Methyl Ester, TRC

Product Code. 30355761
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355761 5 mg 5mg
30362466 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30355761 Supplier Toronto Research Chemicals Supplier No. TRCD2910155MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391054-57-1
Molecular Formula C16H12ClNO3
Molecular Weight (g/mol) 301.72
Synonym 2-(4-Chlorophenyl)-5-benzoxazoleacetic Acid Methyl Ester
SMILES COC(=O)CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H12ClNO3/c1-20-15(19)9-10-2-7-14-13(8-10)18-16(21-14)11-3-5-12(17)6-4-11/h2-8H,9H2,1H3
Chemical Name or Material alpha-Desmethyl Benoxaprofen Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.