missing translation for 'onlineSavingsMsg'
Learn More

Clofedanol Hydrochloride, TRC

Product Code. 30357266
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357266 500 mg -
30356712 1 g 1g
30366742 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357266 Supplier Toronto Research Chemicals Supplier No. TRCC586920500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 511-13-7
Molecular Formula C17 H20 Cl N O . Cl H
Molecular Weight (g/mol) 326.26
Synonym Chlorphedianol Hydrochloride, Clofedanol Hydrochloride, Benzenemethanol, 2-chloro-α-[2-(dimethylamino)ethyl]-α-phenyl-, hydrochloride (1:1), Benzenemethanol, 2-chloro-α-[2-(dimethylamino)ethyl]-α-phenyl-, hydrochloride (9CI), Benzhydrol, 2-chloro-α-[2-(dimethylamino)ethyl]-, hydrochloride (7CI,8CI), 1-o-Chlorophenyl-1-phenyl-3-dimethylamino-1-propanol hydrochloride, 2-Chloro-α-[2-(dimethylamino)ethyl]benzhydrol hydrochloride, Chlophedianol hydrochloride, Clofedanol hydrochloride, Coldrin, Detigon, Pectolitan, Refugal, SK 74, SL 501, ULO, Ulone, α-(2-Dimethylaminoethyl)-o-chlorobenzhydrol hydrochloride
IUPAC Name 1-(2-chlorophenyl)-3-(dimethylamino)-1-phenylpropan-1-ol;hydrochloride
SMILES Cl.CN(C)CCC(O)(c1ccccc1)c2ccccc2Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C17H20ClNO.ClH/c1-19(2)13-12-17(20,14-8-4-3-5-9-14)15-10-6-7-11-16(15)18;/h3-11,20H,12-13H2,1-2H3;1H
Formula Weight 325.1
Percent Purity >95%
Chemical Name or Material Clofedanol Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.