missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopropionaldehyde Diethyl Acetal, TRC

Product Code. 30357380
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357380 25 g 25g
30357379 1 g 1g
30362094 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357380 Supplier Toronto Research Chemicals Supplier No. TRCC98763025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18381-45-8
Molecular Formula C8 H15 N O2
Molecular Weight (g/mol) 157.21
Synonym Butanenitrile, 4,4-diethoxy-, Propionaldehyde, 3-cyano-, diethyl acetal (6CI,7CI), Succinaldehydonitrile, diethyl acetal (8CI), 4,4-Diethoxybutanenitrile, 3-Cyanopropionaldehyde diethyl acetal, 4,4-Diethoxybutyronitrile
IUPAC Name 4,4-diethoxybutanenitrile
SMILES CCOC(CCC#N)OCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H15NO2/c1-3-10-8(11-4-2)6-5-7-9/h8H,3-6H2,1-2H3
Formula Weight 157.1103
Chemical Name or Material 3-Cyanopropionaldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.