missing translation for 'onlineSavingsMsg'
Learn More

Diacetylbenzidine, TRC

Product Code. 30358564
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358564 1 g 1g
30363202 10 g 10g
30367021 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358564 Supplier Toronto Research Chemicals Supplier No. TRCD3091751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-35-4
Molecular Formula C16 H16 N2 O2
Molecular Weight (g/mol) 268.31
IUPAC Name N-[4-(4-acetamidophenyl)phenyl]acetamide
SMILES CC(=O)Nc1ccc(cc1)c2ccc(NC(=O)C)cc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H16N2O2/c1-11(19)17-15-7-3-13(4-8-15)14-5-9-16(10-6-14)18-12(2)20/h3-10H,1-2H3,(H,17,19)(H,18,20)
Formula Weight 268.1212
Chemical Name or Material Diacetylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.