missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloroethyl)-N-methoxy-N,Alpha,Alpha-trimethyl-benzeneacetamide, TRC

Product Code. 30358751
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358751 25 mg 25mg
30349101 2.5 mg 2.5mg
30358437 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358751 Supplier Toronto Research Chemicals Supplier No. TRCC36365025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1638785-17-7
Molecular Formula C14 H20 Cl N O2
Molecular Weight (g/mol) 269.7671
Synonym 4-(2-Chloroethyl)-N-methoxy-N,α,α-trimethylbenzeneacetamide, 2-(4-(3-Chloropropanoyl)phenyl)-N-methoxy-N,2-dimethylpropanamide
IUPAC Name 2-[4-(2-chloroethyl)phenyl]-N-methoxy-N,2-dimethylpropanamide
SMILES CON(C)C(=O)C(C)(C)c1ccc(CCCl)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H20ClNO2/c1-14(2,13(17)16(3)18-4)12-7-5-11(6-8-12)9-10-15/h5-8H,9-10H2,1-4H3
Formula Weight 269.1183
Chemical Name or Material 4-(2-Chloroethyl)-N-methoxy-N,alpha,alpha-trimethyl-benzeneacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.