missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene-d10, TRC

Product Code. 30359540
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359540 1 g 1g
30357378 100 mg 100mg
30364493 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30359540 Supplier Toronto Research Chemicals Supplier No. TRCC9875141G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1603-55-0
Molecular Formula C6 D10
Molecular Weight (g/mol) 92.21
Synonym Cyclohexene-1,2,3,3,4,4,5,5,6,6-d10
IUPAC Name 1,2,3,3,4,4,5,5,6,6-decadeuteriocyclohexene
SMILES [2H]C1=C([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H10/c1-2-4-6-5-3-1/h1-2H,3-6H2/i1D,2D,3D2,4D2,5D2,6D2
Formula Weight 92.141
Chemical Name or Material Cyclohexene-d10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.