missing translation for 'onlineSavingsMsg'
Learn More

Z-Cys(Bzl)-OH, TRC

Product Code. 30360393
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360393 2.5 g 2.5g
30356191 500 mg -
30358218 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360393 Supplier Toronto Research Chemicals Supplier No. TRCC9948002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3257-18-9
Molecular Formula C18H19NO4S
Molecular Weight (g/mol) 345.41
IUPAC Name (2R)-3-benzylsulfanyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OC(=O)[C@H](CSCc1ccccc1)NC(=O)OCc2ccccc2

Specifications

InChI Formula InChI=1S/C18H19NO4S/c20-17(21)16(13-24-12-15-9-5-2-6-10-15)19-18(22)23-11-14-7-3-1-4-8-14/h1-10,16H,11-13H2,(H,19,22)(H,20,21)/t16-/m0/s1
Quantity 2.5 g
Formula Weight 345.1035
Shipping Condition Room Temperature
Chemical Name or Material Z-Cys(Bzl)-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.