missing translation for 'onlineSavingsMsg'
Learn More

22-Desfluoro-22-chlorofluticasone Dimer Impurity, TRC

Product Code. 30362957
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.50mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362957 1 mg 1mg
30360209 0.5 mg 0.50mg
30355063 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362957 Supplier Toronto Research Chemicals Supplier No. TRCD2901051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 22-Desfluoro-22-chlorofluticasone Dimer Impurity
Analyte or Component Names 22-Desfluoro-22-chlorofluticasone Dimer Impurity
Shipping Condition Room Temperature
Molecular Formula C43 H51 Cl F4 O8 S
Synonym (6alpha,11beta,16alpha,17alpha)-6,9-Difluoro-11,17-dihydroxy-16-methyl-3-oxo-Androsta-1,4-diene-17-carboxylic Acid (6alpha,11beta,16alpha,17alpha)-6,9-difluoro-17-[[(chloromethyl)thio]carbonyl]-11-hydroxy-16-methyl-3-oxoandrosta-1,4-dien-17-yl Ester
InChI Formula InChI=1S/C43H51ClF4O8S/c1-20-11-24-26-15-30(45)28-13-22(49)7-9-36(28,3)40(26,47)32(51)17-38(24,5)42(20,55)34(53)56-43(35(54)57-19-44)21(2)12-25-27-16-31(46)29-14-23(50)8-10-37(29,4)41(27,48)33(52)18-39(25,43)6/h7-10,13-14,20-21,24-27,30-33,51-52,55H,11-12,15-19H2,1-6H3/t20-,21-,24+,25+,26+,27+,30+,31+,32+,33-,36+,37+,38+,39-,40+,41+,42+,43+/m1/s1
Solution Type Neat
SMILES C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)O[C@@]5([C@H](C)C[C@H]6[C@@H]7C[C@H](F)C8=CC(=O)C=C[C@]8(C)[C@@]7(F)[C@H](O)C[C@@]56C)C(=O)SCCl
IUPAC Name [(6S,8S,9R,10S,11R,13R,14S,16R,17R)-17-(chloromethylsulfanylcarbonyl)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 839.372
Quantity 1 mg
Formula Weight 838.293
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.