missing translation for 'onlineSavingsMsg'
Learn More

N-[4-Chloro-2-(2-chlorobenzoyl)phenyl]-2-[(2-hydroxyethyl)amino]acetamide, TRC

Product Code. 30365180
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365180 25 mg 25mg
30359597 100 mg -
30363032 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365180 Supplier Toronto Research Chemicals Supplier No. TRCC29300525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24234-92-2
Molecular Formula C17H16Cl2N2O3
Molecular Weight (g/mol) 367.23
Synonym 4'-Chloro-2'-(o-chlorobenzoyl)-2-[(2-hydroxyethyl)amino]acetamide, N-[4-Chloro-2-(2-chlorobenzoyl)phenyl]-2-[(2-hydroxyethyl)amino]acetamide
IUPAC Name N-[4-chloro-2-(2-chlorobenzoyl)phenyl]-2-(2-hydroxyethylamino)acetamide
SMILES OCCNCC(=O)Nc1ccc(Cl)cc1C(=O)c2ccccc2Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H16Cl2N2O3/c18-11-5-6-15(21-16(23)10-20-7-8-22)13(9-11)17(24)12-3-1-2-4-14(12)19/h1-6,9,20,22H,7-8,10H2,(H,21,23)
Formula Weight 366.0538
Chemical Name or Material N-[4-Chloro-2-(2-chlorobenzoyl)phenyl]-2-[(2-hydroxyethyl)amino]acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.