missing translation for 'onlineSavingsMsg'
Learn More

2-Diethylaminoethanol-d10 Hydrochloride, TRC

Product Code. 30370121
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370121 100 mg
30373215 25 mg
30386593 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30370121 Supplier Toronto Research Chemicals Supplier No. TRCD443622100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092978-76-1
Molecular Formula C6H6D10ClNO
Molecular Weight (g/mol) 163.71
Synonym (Diethylamino)ethanol-d10 Hydrochloride, Dehydasal-d10 Hydrochloride, Perdilaton-d10 Hydrochloride
SMILES Cl.[2H]C([2H])([2H])C([2H])([2H])N(CCO)C([2H])([2H])C([2H])([2H])[2H]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H15NO.ClH/c1-3-7(4-2)5-6-8;/h8H,3-6H2,1-2H3;1H/i1D3,2D3,3D2,4D2;
Formula Weight 163.1548
Chemical Name or Material 2-Diethylaminoethanol-d10 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.