missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-5-fluoroindoline-2,3-dione, TRC

Product Code. 30371685
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30371685 50 mg
30373048 100 mg
30369038 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30371685 Supplier Toronto Research Chemicals Supplier No. TRCE94023550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776-47-6
Molecular Formula C10H8NO2F
Molecular Weight (g/mol) 193.17
Synonym 1-Ethyl-5-fluoroindole-2,3-dione, 1-Ethyl-5-fluoro-2,3-dihydro-1H-indole-2,3-dione, 1-Ethyl-5-fluoro-1H-indole-2,3-dione, Isatin, 1-ethyl-5-fluoro- (6CI), Indole-2,3-dione, 1-ethyl-5-fluoro- (7CI,8CI)
SMILES CCN1C(=O)C(=O)c2cc(F)ccc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H8FNO2/c1-2-12-8-4-3-6(11)5-7(8)9(13)10(12)14/h3-5H,2H2,1H3
Formula Weight 193.0539
Chemical Name or Material 1-Ethyl-5-fluoroindoline-2,3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.