missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Hydroxyphenylacetate Trifluoromethanesulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-Hydroxyphenylacetate Trifluoromethanesulfonate |
| Analyte or Component Names | Ethyl 2-Hydroxyphenylacetate Trifluoromethanesulfonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H11 F3 O5 S |
| InChI Formula | InChI=1S/C11H11F3O5S/c1-2-18-10(15)7-8-5-3-4-6-9(8)19-20(16,17)11(12,13)14/h3-6H,2,7H2,1H3 |
| Solution Type | Neat |
| SMILES | CCOC(=O)Cc1ccccc1OS(=O)(=O)C(F)(F)F |
| IUPAC Name | ethyl 2-[2-(trifluoromethylsulfonyloxy)phenyl]acetate |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 312.262 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?