missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Arachidonate, TRC

Product Code. 30379869
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379869 1 g
30388387 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379869 Supplier Toronto Research Chemicals Supplier No. TRCE8994001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1808-26-0
Molecular Formula C22 H36 O2
Molecular Weight (g/mol) 332.52
Synonym (5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraenoic Acid Ethyl Ester, (all-Z)-5,8,11,14-Eicosatetraenoic Acid Ethyl Ester, Arachidonic Acid Ethyl Ester
IUPAC Name ethyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
SMILES CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24-4-2/h8-9,11-12,14-15,17-18H,3-7,10,13,16,19-21H2,1-2H3/b9-8-,12-11-,15-14-,18-17-
Formula Weight 332.2715
Chemical Name or Material Ethyl Arachidonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.