missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2-Methoxy-2-oxoethyl) Malonate, TRC

Product Code. 30388619
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388619 500 mg 500mg
30381505 1 g 1g
30384238 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388619 Supplier Toronto Research Chemicals Supplier No. TRCE572465500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl (2-Methoxy-2-oxoethyl) Malonate
Analyte or Component Names Ethyl (2-Methoxy-2-oxoethyl) Malonate
Shipping Condition Room Temperature
Molecular Formula C8 H12 O6
InChI Formula InChI=1S/C8H12O6/c1-3-13-6(9)4-7(10)14-5-8(11)12-2/h3-5H2,1-2H3
Solution Type Neat
SMILES CCOC(=O)CC(=O)OCC(=O)OC
IUPAC Name 1-O-ethyl 3-O-(2-methoxy-2-oxoethyl) propanedioate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 204.177
Quantity 500 mg
Formula Weight 204.063
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.