missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxy Nortriptyline Maleate, TRC

Product Code. 30389534
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389534 2.5 mg 2.50mg
30399462 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389534 Supplier Toronto Research Chemicals Supplier No. TRCH9488202.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74853-74-0
Molecular Formula C23H25NO
Molecular Weight (g/mol) 395.45
Synonym (5E)-10,11-Dihydro-5-[3-(methylamino)propylidene]-5H-dibenzo[a,d]cyclohepten-10-ol (2Z)-2-Butenedioate, (E)-10-Hydroxynortriptyline Maleate, 10-Hydroxy-(E)-nortriptyline Maleate, trans-10-Hydroxynortriptyline Maleate
SMILES CNCC/C=C/1\C2=CC=CC=C2CC(C3=CC=CC=C13)O.C(=C/C(=O)O)/C(=O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C19H21NO/c1-20-12-6-11-16-15-8-3-2-7-14(15)13-19(21)18-10-5-4-9-17(16)18/h2-5,7-11,19-21H,6,12-13H2,1H3/b16-11+
Formula Weight 395.1733
Chemical Name or Material 10-Hydroxy Nortriptyline Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.