missing translation for 'onlineSavingsMsg'
Learn More

2-Heptacosanone, TRC

Product Code. 30391991
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391991 250 mg
30407627 50 mg
30407965 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391991 Supplier Toronto Research Chemicals Supplier No. TRCH265240250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7796-19-2
Molecular Formula C27H54O
Molecular Weight (g/mol) 394.72
Synonym Heptacosan-2-one
IUPAC Name heptacosan-2-one
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C27H54O/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27(2)28/h3-26H2,1-2H3
Formula Weight 394.4175
Chemical Name or Material 2-Heptacosanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.