missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic Acid-L-glycine-L-glycine-L-isoleucine-L-tryptophan-L-phenylalanine-L-proline-L-tyrosine-L-glycine-L-glycine-L-phenylalanine TFA Salt (95%), TRC
Specifications
Specifications
| Chemical Name or Material | L-Glutamic Acid-L-glycine-L-glycine-L-isoleucine-L-tryptophan-L-phenylalanine-L-proline-L-tyrosine-L-glycine-L-glycine-L-phenylalanine TFA Salt |
| Analyte or Component Names | L-Glutamic Acid-L-glycine-L-glycine-L-isoleucine-L-tryptophan-L-phenylalanine-L-proline-L-tyrosine-L-glycine-L-glycine-L-phenylalanine TFA Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C62H76N12O15 •(C2HF3O2) |
| Synonym | 5-((1H-Indol-3-yl)methyl)-17-amino-2-benzyl-8-(sec-butyl)-1-(2-((1-((2-((2-((1-carboxy-2-phenylethyl)amino)-2-oxoethyl)amino)-2-oxoethyl)amino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)carbamoyl)pyrrolidin-1-yl)-1,4,7,10,13,16-hexaoxo-3,6,9,12,15-pentaazaicosan-20-oic Acid Trifluoroacetic Acid Salt |
| InChI Formula | InChI=1S/C62H76N12O15.C2HF3O/c1-3-37(2)56(72-54(80)34-65-52(78)35-67-73-42(36-75)22-25-55(81)82)60(86)70-47(30-41-31-63-45-18-11-10-17-44(41)45)58(84)71-48(28-38-13-6-4-7-14-38)61(87)74-26-12-19-50(74)59(85)69-46(27-40-20-23-43(76)24-21-40)57(83)66-32-51(77)64-33-53(79)68-49(62(88)89)29-39-15-8-5-9-16-39;3-2(4,5)1-6/h4-11,13-18,20-21,23-24,31,36-37,42,46-50,56,63,67,73,76H,3,12,19,22,25-30,32-35H2,1-2H3,(H,64,77)(H,65,78)(H,66,83)(H,68,79)(H,69,85)(H,70,86)(H,71,84)(H,72,80)(H,81,82)(H,88,89);1H/t37-,42-,46-,47-,48-,49-,50-,56-;/m0./s1 |
| Solution Type | Neat |
| SMILES | OC([C@H](CC1=CC=CC=C1)NC(CNC(CNC([C@H](CC2=CC=C(C=C2)O)NC([C@H]3N(C([C@H](CC4=CC=CC=C4)NC([C@H](CC5=CNC6=C5C=CC=C6)NC([C@H]([C@H](CC)C)NC(CNC(CNN[C@H](C([H])=O)CCC(O)=O)=O)=O)=O)=O)=O)CCC3)=O)=O)=O)=O)=O.O=CC(F)(F)F |
| IUPAC Name | (2S,5S,8S,17S)-5-((1H-indol-3-yl)methyl)-2-benzyl-8-((S)-sec-butyl)-1-((S)-2-(((S)-1-((2-((2-(((S)-1-carboxy-2-phenylethyl)amino)-2-oxoethyl)amino)-2-oxoethyl)amino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)carbamoyl)pyrrolidin-1-yl)-17-formyl-1,4,7,10,13-pentaoxo-3,6,9,12,15,16-hexaazaicosan-20-oic acid 2,2,2-trifluoro-1l3-ethan-1-one |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?