missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isosteviol Acyl-Beta-D-glucuronide Potassium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Isosteviol Acyl-beta-D-glucuronide Potassium Salt |
| Analyte or Component Names | Isosteviol Acyl-beta-D-glucuronide Potassium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26H37KO9 |
| Synonym | 1-[(4alpha)-13-Methyl-16-oxo-17-norkauran-18-oate]-beta-D-glucopyranuronic Acid Potassium Salt, Isosteviol GlucI902215uronide Potassium Salt |
| InChI Formula | InChI=1S/C26H38O9.K/c1-23-9-5-14-24(2)7-4-8-25(3,13(24)6-10-26(14,12-23)11-15(23)27)22(33)35-21-18(30)16(28)17(29)19(34-21)20(31)32;/h13-14,16-19,21,28-30H,4-12H2,1-3H3,(H,31,32);/q;+1/p-1/t13-,14-,16-,17-,18+,19-,21-,23-,24+,25+,26-;/m0./s1 |
| Solution Type | Neat |
| SMILES | [K+].C[C@@]12CC[C@@H]3[C@@](CC[C@H]4[C@@]3(C)CCC[C@@]4(C)C(=O)O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)[O-])(CC1=O)C2 |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 532.66 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?