missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Hydroxy-4-Methyl Coumarin Glucuronide Cyclohexanamine Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 7-Hydroxy-4-Methyl Coumarin Glucuronide Cyclohexanamine Salt |
| Analyte or Component Names | 7-Hydroxy-4-Methyl Coumarin Glucuronide Cyclohexanamine Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H16 O9 . C6 H13 N |
| Synonym | (2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-((5-methyl-7-oxo-7,8-dihydronaphthalen-2-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid cyclohexamine |
| InChI Formula | InChI=1S/C16H16O9.C6H13N/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22;7-6-4-2-1-3-5-6/h2-5,11-14,16,18-20H,1H3,(H,21,22);6H,1-5,7H2/t11-,12-,13+,14-,16+;/m0./s1 |
| Solution Type | Neat |
| SMILES | CC1=CC(=O)Oc2cc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)ccc12.NC4CCCCC4 |
| IUPAC Name | cyclohexanamine;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?