missing translation for 'onlineSavingsMsg'
Learn More

1H,1aH,6H,6aH-cyclopropa[a]indene-1-carboxylic acid, TRC

Product Code. 30406814
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406814 2.5 mg 2.5mg
30405174 25 mg 25mg
30408331 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406814 Supplier Toronto Research Chemicals Supplier No. TRCH9554332.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501418-06-0
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.2
IUPAC Name 1,1a,6,6a-tetrahydrocyclopropa[a]indene-1-carboxylic acid
SMILES OC(=O)C1C2Cc3ccccc3C12

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C11H10O2/c12-11(13)10-8-5-6-3-1-2-4-7(6)9(8)10/h1-4,8-10H,5H2,(H,12,13)
Formula Weight 174.0681
Chemical Name or Material 1H,1aH,6H,6aH-cyclopropa[a]indene-1-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.