missing translation for 'onlineSavingsMsg'
Learn More

Octahydroindoline-2-carboxylic Acid Benzyl Ester (-)-Dibenzoyl-L-tartaric Acid, TRC

Product Code. 30409360
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409360 50 mg 50mg
30416551 1 g 1g
30427622 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409360 Supplier Toronto Research Chemicals Supplier No. TRCO23650550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octahydroindoline-2-carboxylic Acid Benzyl Ester (-)-Dibenzoyl-L-tartaric Acid
Analyte or Component Names Octahydroindoline-2-carboxylic Acid Benzyl Ester (-)-Dibenzoyl-L-tartaric Acid
Shipping Condition Room Temperature
Molecular Formula C34H35NO10
InChI Formula InChI=1S/C18H14O8.C16H21NO2/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12;18-16(19-11-12-6-2-1-3-7-12)15-10-13-8-4-5-9-14(13)17-15/h1-10,13-14H,(H,19,20)(H,21,22);1-3,6-7,13-15,17H,4-5,8-11H2
Solution Type Neat
SMILES O=C(C1CC2C(CCCC2)N1)OCC3=CC=CC=C3.O=C([C@H](OC(C4=CC=CC=C4)=O)[C@H](C(O)=O)OC(C5=CC=CC=C5)=O)O
IUPAC Name benzyl octahydro-1H-indole-2-carboxylate (2R,3R)-2,3-bis(benzoyloxy)succinate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 617.64
Quantity 50 mg
Formula Weight 617.23
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.