missing translation for 'onlineSavingsMsg'
Learn More

Sumatriptan-d6 Succinate, TRC

Product Code. 30411152
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411152 10 mg -
30426968 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30411152 Supplier Toronto Research Chemicals Supplier No. TRCS81000210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215621-31-0
Molecular Formula C14 2H6 H15 N3 O2 S . C4 H6 O4
Molecular Weight (g/mol) 419.53
Synonym 1-[3-[2-(Bis(trideuteriomethyl)amino)ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide butanedioic acid, 3-[2-[Di(methyl-d3)amino]ethyl]-N-methyl-1H-indole-5-methanesulfonamide 1,4-butanedioate (1:1), 1-[3-[2-[Di(methyl-d3)amino]ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide succinate, Sumatriptan-d6 succinate, Sumatriptan-D6 (N,N-dimethylamino-D6) Succinate
IUPAC Name 1-[3-[2-[bis(trideuteriomethyl)amino]ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide;butanedioic acid
SMILES [2H]C([2H])([2H])N(CCc1c[nH]c2ccc(CS(=O)(=O)NC)cc12)C([2H])([2H])[2H].OC(=O)CCC(=O)O

Specifica

Quantity 10 mg
InChI Formula InChI=1S/C14H21N3O2S.C4H6O4/c1-15-20(18,19)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2)3;5-3(6)1-2-4(7)8/h4-5,8-9,15-16H,6-7,10H2,1-3H3;1-2H2,(H,5,6)(H,7,8)/i2D3,3D3;
Formula Weight 419.1997
Chemical Name or Material Sumatriptan-d6 Succinate
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.