missing translation for 'onlineSavingsMsg'
Learn More

SL0101, TRC

Product Code. 30412989
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30412989 50 mg
30425806 10 mg
30423586 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412989 Supplier Toronto Research Chemicals Supplier No. TRCS56000050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77307-50-7
Molecular Formula C25 H24 O12
Molecular Weight (g/mol) 516.45
Synonym 4H-1-Benzopyran-4-one, 3-[(3,4-di-O-acetyl-6-deoxy-α-L-mannopyranosyl)oxy]-5,7-dihydroxy-2-(4-hydroxyphenyl)-, 3-[(3,4-Di-O-acetyl-6-deoxy-α-L-mannopyranosyl)oxy]-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 3' ',4' '-Di-O-acetylafzelin, Kaempferol 3-O-(3' ',4' '-di-O-acetyl-α-L-rhamnopyranoside), SL 0101, SL 0101-1
IUPAC Name [(2S,3S,4S,5R,6S)-4-acetyloxy-6-[5,7-dihydroxy-2-(4-hydroxyphenyl)-4-oxochromen-3-yl]oxy-5-hydroxy-2-methyloxan-3-yl] acetate
SMILES C[C@@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)cc4)[C@H](O)[C@H](OC(=O)C)[C@H]1OC(=O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C25H24O12/c1-10-21(34-11(2)26)24(35-12(3)27)20(32)25(33-10)37-23-19(31)18-16(30)8-15(29)9-17(18)36-22(23)13-4-6-14(28)7-5-13/h4-10,20-21,24-25,28-30,32H,1-3H3/t10-,20+,21-,24-,25-/m0/s1
Formula Weight 516.1268
Percent Purity >95%
Chemical Name or Material SL0101
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.