missing translation for 'onlineSavingsMsg'
Learn More

Oxatomide, TRC

Product Code. 30414244
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414244 10 mg
30426626 100 mg
30417418 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30414244 Supplier Toronto Research Chemicals Supplier No. TRCO84510010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60607-34-3
Molecular Formula C27 H30 N4 O
Molecular Weight (g/mol) 426.55
Synonym 2H-Benzimidazol-2-one, 1-[3-[4-(diphenylmethyl)-1-piperazinyl]propyl]-1,3-dihydro-, Celtect, Cobiona, Dasten, KW 4354, NSC 309710, Oxatomide, R 35443, Tinset
IUPAC Name 3-[3-(4-benzhydrylpiperazin-1-yl)propyl]-1H-benzimidazol-2-one
SMILES O=C1Nc2ccccc2N1CCCN3CCN(CC3)C(c4ccccc4)c5ccccc5

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H30N4O/c32-27-28-24-14-7-8-15-25(24)31(27)17-9-16-29-18-20-30(21-19-29)26(22-10-3-1-4-11-22)23-12-5-2-6-13-23/h1-8,10-15,26H,9,16-21H2,(H,28,32)
Formula Weight 426.242
Chemical Name or Material Oxatomide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.