missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-O-Allyl Rocuronium Bromide (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 3-O-Allyl Rocuronium Bromide |
| Analyte or Component Names | 3-O-Allyl Rocuronium Bromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C35 H57 N2 O4 . Br |
| Synonym | 3-Propenoxy Rocuronium Bromide, Rocuronium Bromide Impurity 1 |
| InChI Formula | InChI=1S/C35H57N2O4.BrH/c1-6-16-37(17-8-9-18-37)31-23-29-27-11-10-26-22-32(40-19-7-2)30(36-14-20-39-21-15-36)24-35(26,5)28(27)12-13-34(29,4)33(31)41-25(3)38;/h6-7,26-33H,1-2,8-24H2,3-5H3;1H/q+1;/p-1/t26-,27+,28-,29-,30-,31-,32-,33-,34-,35-;/m0./s1 |
| Solution Type | Neat |
| SMILES | [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](OCC=C)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N5CCOCC5)[N+]6(CC=C)CCCC6 |
| IUPAC Name | [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-3-allyloxy-16-(1-allylpyrrolidin-1-ium-1-yl)-10,13-dimethyl-2-morpholino-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate;bromide |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?