missing translation for 'onlineSavingsMsg'
Learn More

Orphenadrine Citrate Salt, TRC

Product Code. 30416119
Change view
Click to view available options
Quantity:
50 mg
5 g
10 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416119 5 g -
30422427 50 mg -
30425389 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416119 Supplier Toronto Research Chemicals Supplier No. TRCO6953005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4682-36-4
Molecular Formula C18 H23 N O . C6 H8 O7
Molecular Weight (g/mol) 461.5
Synonym Orphenadrine citrate, (RS)-N,N-Dimethyl-2-[(2-methylphenyl)phenylmethoxy]ethanamine dihydrogen 2-hydroxypropane-1,2,3-tricarboxylate
IUPAC Name N,N-dimethyl-2-[(2-methylphenyl)-phenylmethoxy]ethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CN(C)CCOC(c1ccccc1)c2ccccc2C.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C18H23NO.C6H8O7/c1-15-9-7-8-12-17(15)18(20-14-13-19(2)3)16-10-5-4-6-11-16;7-3(8)1-6(13,5(11)12)2-4(9)10/h4-12,18H,13-14H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Formula Weight 461.205
Percent Purity >95%
Chemical Name or Material Orphenadrine citrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.