missing translation for 'onlineSavingsMsg'
Learn More

7-Oxo Cholesterol 3-Acetate, TRC

Product Code. 30416807
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30416807 25 mg
30414989 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30416807 Supplier Toronto Research Chemicals Supplier No. TRCO86998525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 809-51-8
Molecular Formula C29H46O3
Molecular Weight (g/mol) 442.68
Synonym (3beta)-3-(Acetyloxy)cholest-5-en-7-one, 3beta-Hydroxy-cholest-5-en-7-one Acetate, 7-Ketocholesteryl Acetate, 7-Oxocholest-5-en-3beta-yl Acetate, 7-Oxocholesteryl Acetate, NSC 59469
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-7-oxo-1,2,3,4,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-yl] acetate
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@H]3[C@H](CC[C@]12C)[C@@]4(C)CC[C@@H](CC4=CC3=O)OC(=O)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C29H46O3/c1-18(2)8-7-9-19(3)23-10-11-24-27-25(13-15-29(23,24)6)28(5)14-12-22(32-20(4)30)16-21(28)17-26(27)31/h17-19,22-25,27H,7-16H2,1-6H3/t19-,22+,23-,24+,25+,27+,28+,29-/m1/s1
Formula Weight 442.3447
Chemical Name or Material 7-Oxo Cholesterol 3-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.