missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puromycin Aminonucleoside Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Puromycin Aminonucleoside Hydrochloride |
| Analyte or Component Names | Puromycin Aminonucleoside Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12H18N6O3 • HCl |
| Synonym | 3'-Amino-3'-deoxy-N,N-dimethyladenosine Hydrochloride, 3'-Amino-3'-deoxy-N6,N6-dimethyladenosine Hydrochloride, 6-(Dimethylamino)-9-(3-amino-3-deoxy-β-D-ribofuranosyl)purine Hydrochloride, 6-Dimethylamino-9-(3-amino-3-deoxyribosyl)purine Hydrochloride, 9-(3-Amino-3-deoxy-β-D-ribofuranosyl)-6-(dimethylamino)-9H-purine Hydrochloride, Aminonucleoside Hydrochloride, Aminonucleoside Puromycin Hydrochloride, NSC 3056 Hydrochloride, PANS Hydrochloride, Aminonucleoside Puromycin Hydrochloride, SAN Hydrochloride, Stylomycin Aminonucleoside Hydrochloride |
| InChI Formula | InChI=1S/C12H18N6O3.ClH/c1-17(2)10-8-11(15-4-14-10)18(5-16-8)12-9(20)7(13)6(3-19)21-12;/h4-7,9,12,19-20H,3,13H2,1-2H3;1H/t6-,7-,9-,12-;/m1./s1 |
| Solution Type | Neat |
| SMILES | OC[C@@H]1[C@@H](N)[C@@H](O)[C@H](N2C(N=CN=C3N(C)C)=C3N=C2)O1.Cl |
| IUPAC Name | (2R,3R,4S,5S)-4-amino-2-(6-(dimethylamino)-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3-ol hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?