missing translation for 'onlineSavingsMsg'
Learn More

SR 2640 Hydrochloride, TRC

Product Code. 30419583
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30419583 50 mg
30426617 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30419583 Supplier Toronto Research Chemicals Supplier No. TRCS68451050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146662-42-2
Molecular Formula C23H18N2O3 . HCl
Molecular Weight (g/mol) 406.9
Synonym 2-[[3-(2-Quinolinylmethoxy)phenyl]amino]Benzoic Acid Monohydrochloride (9CI), SR 2640 Hydrochloride
IUPAC Name 2-[3-(quinolin-2-ylmethoxy)anilino]benzoic acid;hydrochloride
SMILES Cl.OC(=O)c1ccccc1Nc2cccc(OCc3ccc4ccccc4n3)c2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H18N2O3.ClH/c26-23(27)20-9-2-4-11-22(20)24-17-7-5-8-19(14-17)28-15-18-13-12-16-6-1-3-10-21(16)25-18;/h1-14,24H,15H2,(H,26,27);1H
Formula Weight 406.1084
Chemical Name or Material 2-((3-(2-Quinolinylmethoxy)phenyl)amino)Benzoic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.