missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl 5Beta-Dihydrocortisone 1,3-Dioxane, TRC

Product Code. 30419943
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419943 10 mg 10mg
30425717 25 mg 25mg
30423757 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419943 Supplier Toronto Research Chemicals Supplier No. TRCP31936010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Phenyl 5beta-Dihydrocortisone 1,3-Dioxane
Analyte or Component Names 2-Phenyl 5beta-Dihydrocortisone 1,3-Dioxane
Shipping Condition Room Temperature
Molecular Formula C28H34O5
Synonym (4’R,5R,8S,9S,10S,13S,14S)-10,13-Dimethyl-2’-phenyldodecahydrospiro[cyclopenta[a]phenanthrene-17,4’-[1,3]dioxane]-3,5’11(2H,4H)-trione
InChI Formula InChI=1S/C28H34O5/c1-26-12-10-19(29)14-18(26)8-9-20-21-11-13-28(27(21,2)15-22(30)24(20)26)23(31)16-32-25(33-28)17-6-4-3-5-7-17/h3-7,18,20-21,24-25H,8-16H2,1-2H3/t18-,20+,21+,24-,25?,26+,27+,28+/m1/s1
Solution Type Neat
SMILES O=C1CC[C@@]2(C)[C@@](CC[C@]([C@@](CC[C@]34C(COC(C5=CC=CC=C5)O4)=O)([H])[C@]3(C)C6)([H])[C@]2([H])C6=O)([H])C1
IUPAC Name (5R,8S,9S,10S,13S,14S,17R)-10,13-dimethyl-2'-phenyltetradecahydrospiro[cyclopenta[a]phenanthrene-17,4'-[1,3]dioxane]-3,5',11-trione
Industry Type Pharmaceutical
Molecular Weight (g/mol) 450.57
Quantity 10 mg
Formula Weight 450.24
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.