missing translation for 'onlineSavingsMsg'
Learn More

1-O-Octadecyl-2-O-benzyl-rac-glycerol, TRC

Product Code. 30423443
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423443 1 g -
30420753 100 mg -
30416572 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423443 Supplier Toronto Research Chemicals Supplier No. TRCO2359501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86008-21-1
Molecular Formula C28H50O3
Molecular Weight (g/mol) 434.69
Synonym 3-(Octadecyloxy)-2-(phenylmethoxy)-1-propanol, 2-(Benzyloxy)-3-(octadecyloxy)propan-1-ol
IUPAC Name 3-octadecoxy-2-phenylmethoxypropan-1-ol
SMILES CCCCCCCCCCCCCCCCCCOCC(CO)OCc1ccccc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C28H50O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20-23-30-26-28(24-29)31-25-27-21-18-17-19-22-27/h17-19,21-22,28-29H,2-16,20,23-26H2,1H3
Formula Weight 434.376
Chemical Name or Material 1-O-Octadecyl-2-O-benzyl-rac-glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.