missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ticagrelor TCG Sulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | Ticagrelor TCG Sulfonate |
| Analyte or Component Names | Ticagrelor TCG Sulfonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H28 F2 N6 O7 S2 |
| Synonym | 2-(((1S,2S,3S,4R)-4-(7-(((1R,2S)-2-(3,4-difluorophenyl)cyclopropyl)amino)-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)-2,3-dihydroxycyclopentyl)oxy)ethyl hydrogen sulfate |
| InChI Formula | InChI=1S/C23H28F2N6O7S2/c1-2-7-39-23-27-21(26-15-9-12(15)11-3-4-13(24)14(25)8-11)18-22(28-23)31(30-29-18)16-10-17(20(33)19(16)32)37-5-6-38-40(34,35)36/h3-4,8,12,15-17,19-20,32-33H,2,5-7,9-10H2,1H3,(H,26,27,28)(H,34,35,36)/t12-,15+,16+,17-,19-,20+/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCSc1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](OCCOS(=O)(=O)O)[C@@H](O)[C@H]5O)c4n1 |
| IUPAC Name | 2-[(1S,2S,3S,4R)-4-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-propylsulfanyl-triazolo[4,5-d]pyrimidin-3-yl]-2,3-dihydroxy-cyclopentoxy]ethyl hydrogen sulfate |
| Molecular Weight (g/mol) | 602.63 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?