missing translation for 'onlineSavingsMsg'
Learn More

L-Xylo-2-Hexulosonic Acid Hydrate, TRC

Product Code. 30453046
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453046 25 g 25g
30450113 50 g 50g
30452714 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453046 Supplier Toronto Research Chemicals Supplier No. TRCX75030525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 342385-52-8
Molecular Formula C6 H10 O7 . H2 O
Molecular Weight (g/mol) 212.155
Synonym 2-Keto-L-gulonic Acid Hydrate, 2-Oxo-gulonic Acid Hydrate, Provitamin C Hydrate, D-Sorbosonic Acid Hydrate
IUPAC Name (3S,4R,5S)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid;hydrate
SMILES O.OC[C@H](O)[C@@H](O)[C@H](O)C(=O)C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H10O7.H2O/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-4,7-10H,1H2,(H,12,13);1H2/t2-,3+,4-;/m0./s1
Formula Weight 212.053
Percent Purity >95%
Chemical Name or Material L-Xylo-2-Hexulosonic Acid Hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.