missing translation for 'onlineSavingsMsg'
Learn More

[6-(Trimethylammonium)hexyl] Methanethiosulfonate Bromide, TRC

Product Code. 30454519
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454519 10 mg -
30454550 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454519 Supplier Toronto Research Chemicals Supplier No. TRCT79590710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1041424-77-4
Molecular Formula C10 H24 N O2 S2 . Br
Molecular Weight (g/mol) 334.34
IUPAC Name trimethyl(6-methylsulfonylsulfanylhexyl)azanium;bromide
SMILES [Br-].C[N+](C)(C)CCCCCCSS(=O)(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H24NO2S2.BrH/c1-11(2,3)9-7-5-6-8-10-14-15(4,12)13;/h5-10H2,1-4H3;1H/q+1;/p-1
Formula Weight 333.0432
Chemical Name or Material (6-(Trimethylammonium)hexyl) Methanethiosulfonate Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.