missing translation for 'onlineSavingsMsg'
Learn More

1,4,7-Trioxacyclohexadecane-8,16-dione, TRC

Product Code. 30455451
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455451 250 mg 250mg
30456714 50 mg 50mg
30455578 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455451 Supplier Toronto Research Chemicals Supplier No. TRCT767275250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70685-15-3
Molecular Formula C13H22O5
Molecular Weight (g/mol) 258.31
IUPAC Name 1,4,7-trioxacyclohexadecane-8,16-dione
SMILES O=C1CCCCCCCC(=O)OCCOCCO1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H22O5/c14-12-6-4-2-1-3-5-7-13(15)18-11-9-16-8-10-17-12/h1-11H2
Formula Weight 258.1467
Chemical Name or Material 1,4,7-Trioxacyclohexadecane-8,16-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.