missing translation for 'onlineSavingsMsg'
Learn More

Methapyrilene-d6 Hydrochloride, TRC

Product Code. 30464383
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464383 1 mg 1mg
30472031 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30464383 Supplier Toronto Research Chemicals Supplier No. TRCM2599721MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2985926-49-4
Molecular Formula C14H13D6N3S . HCl
Molecular Weight (g/mol) 303.88
Synonym 2-[[2-(Dimethylamino)ethyl]-2-thenylamino]pyridine-d6 hydrochloride, Thenylene-d6 hydrochloride, Thenylpyramine-d6 hydrochloride
IUPAC Name N1,N1-bis(methyl-d3)-N2-(pyridin-2-yl)-N2-(thiophen-2-ylmethyl)ethane-1,2-diamine hydrochloride
SMILES [2H]C([2H])([2H])N(C([2H])([2H])[2H])CCN(CC1=CC=CS1)C2=NC=CC=C2.Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H19N3S.ClH/c1-16(2)9-10-17(12-13-6-5-11-18-13)14-7-3-4-8-15-14;/h3-8,11H,9-10,12H2,1-2H3;1H/i1D3,2D3;
Formula Weight 303.15
Chemical Name or Material Methapyrilene-d6 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.