missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyridin-2-one Pirfenidone, TRC

Product Code. 30465457
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465457 100 mg 100mg
30471917 10 mg 10mg
30474317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30465457 Supplier Toronto Research Chemicals Supplier No. TRCM493285100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2251047-36-4
Molecular Formula C18H16N2O2
Molecular Weight (g/mol) 292.33
Synonym 1,1'-(1,4-Phenylene)bis[5-methyl-2(1H)-pyridinone
IUPAC Name 5-methyl-1-[4-(5-methyl-2-oxopyridin-1-yl)phenyl]pyridin-2-one
SMILES CC1=CN(C(=O)C=C1)c2ccc(cc2)N3C=C(C)C=CC3=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H16N2O2/c1-13-3-9-17(21)19(11-13)15-5-7-16(8-6-15)20-12-14(2)4-10-18(20)22/h3-12H,1-2H3
Formula Weight 292.1212
Chemical Name or Material 5-Methylpyridin-2-one Pirfenidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.